accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 484-17-3

HO

CAS RN 484-17-3

IUPAC Name: phenanthren-9-ol
Synonyms: 9-phenanthrol
Molecular Weight (g/mol): 194.23
Molecular Formula: C14H10O
InChi Key: DZKIUEHLEXLYKM-UHFFFAOYSA-N
SMILES: OC1=CC2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results