CAS RN 484-29-7
CAS RN 484-29-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dictamnine, TRC
CAS: 484-29-7 Molecular Formula: C12H9NO2 Molecular Weight (g/mol): 199.21 Synonym: Dictamnine,4-Methoxyfuro[2,3-b]quinoline,Dectamine,Dictamine SMILES: COC1=C2C(OC=C2)=NC3=CC=CC=C13