accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

N(S)(R)(R)NH2CHOHH

CAS RN 485-71-2

IUPAC Name: {5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl}(quinolin-4-yl)methanol
Synonyms: cinchonidine sulfate cinchonine sulfate
Molecular Weight (g/mol): 294.4
Molecular Formula: C19H22N2O
InChi Key: KMPWYEUPVWOPIM-UHFFFAOYNA-N
SMILES: OC(C1CC2CCN1CC2C=C)C1=C2C=CC=CC2=NC=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)