accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClHNN

CAS RN 4857-04-9

IUPAC Name: 2-(chloromethyl)-1H-1,3-benzodiazole
Synonyms: 2-(chloromethyl)-1H-1,3-benzodiazole
Molecular Weight (g/mol): 166.61
Molecular Formula: C8H7ClN2
InChi Key: SPMLMLQATWNZEE-UHFFFAOYSA-N
SMILES: ClCC1=NC2=CC=CC=C2N1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results