accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2FCl

CAS RN 4863-91-6

IUPAC Name: 3-chloro-5-fluoroaniline
Synonyms: 3-chloro-5-fluoroaniline
Molecular Weight (g/mol): 145.56
Molecular Formula: C6H5ClFN
InChi Key: LPIFAHAICWJMRR-UHFFFAOYSA-N
SMILES: NC1=CC(F)=CC(Cl)=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results