accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 486414-83-9

IUPAC Name: 1-(1-methyl-1H-imidazol-4-yl)methanamine
Synonyms: 1-(1-methylimidazol-4-yl)methanamine
Molecular Weight (g/mol): 111.15
Molecular Formula: C5H9N3
InChi Key: YSEAGFBRAQOCFM-UHFFFAOYSA-N
SMILES: CN1C=NC(CN)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results