accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 487-26-3

IUPAC Name: 2-phenyl-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms: flavanone, (+-)- (r)-flavanone (+-)-isomer flavanone flavanone, 2-(14)C-labeled flavanone, (+-)-isomer 2-(14)c-labeled flavanone 2-(14)C-labeled flavanone (r)-isomer flavanone (S)-flavanone flavanone, (s)-isomer
Molecular Weight (g/mol): 224.26
Molecular Formula: C15H12O2
InChi Key: ZONYXWQDUYMKFB-UHFFFAOYNA-N
SMILES: O=C1CC(OC2=CC=CC=C12)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results