accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4877-80-9

IUPAC Name: triphenylene-2,3,6,7,10,11-hexol
Synonyms: triphenylene-2,3,6,7,10,11-hexol
Molecular Weight (g/mol): 324.29
Molecular Formula: C18H12O6
InChi Key: QMLILIIMKSKLES-UHFFFAOYSA-N
SMILES: OC1=C(O)C=C2C(=C1)C1=CC(O)=C(O)C=C1C1=CC(O)=C(O)C=C21

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results