accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 488-17-5

CH3HOHO

CAS RN 488-17-5

IUPAC Name: 3-methylbenzene-1,2-diol
Synonyms: 3-methyl-benzene-1,2-diol dihydroxytoluene
Molecular Weight (g/mol): 124.14
Molecular Formula: C7H8O2
InChi Key: PGSWEKYNAOWQDF-UHFFFAOYSA-N
SMILES: CC1=CC=CC(O)=C1O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)