accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 489-84-9

IUPAC Name: 1,4-dimethyl-7-(propan-2-yl)azulene
Synonyms: azulon guaiazulene vetivazulen S-guaiazulene azulen-beris
Molecular Weight (g/mol): 198.31
Molecular Formula: C15H18
InChi Key: FWKQNCXZGNBPFD-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(C)C2=CC=C(C)C2=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)