accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONHN

CAS RN 491-36-1

IUPAC Name: 1,4-dihydroquinazolin-4-one
Synonyms: quinazolinone
Molecular Weight (g/mol): 146.15
Molecular Formula: C8H6N2O
InChi Key: QMNUDYFKZYBWQX-UHFFFAOYSA-N
SMILES: O=C1N=CNC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results