accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOHOOOHOH

CAS RN 491-70-3

IUPAC Name: 2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-4H-chromen-4-one
Synonyms: luteolin C.I. natural yellow 2 weld lake yama kariyasu cyanidenon 1470
Molecular Weight (g/mol): 286.24
Molecular Formula: C15H10O6
InChi Key: IQPNAANSBPBGFQ-UHFFFAOYSA-N
SMILES: OC1=CC(O)=C2C(=O)C=C(OC2=C1)C1=CC=C(O)C(O)=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)