accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4911-54-0

H3COOCH2CH3

CAS RN 4911-54-0

IUPAC Name: ethyl 4-methylpent-4-enoate
Synonyms: ethyl 4-methylpent-4-enoate
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: RRTBSPBUHUUTHR-UHFFFAOYSA-N
SMILES: CCOC(=O)CCC(C)=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results