accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClS

CAS RN 4911-65-3

IUPAC Name: [(3-chloropropyl)sulfanyl]benzene
Synonyms: [(3-chloropropyl)sulfanyl]benzene
Molecular Weight (g/mol): 186.7
Molecular Formula: C9H11ClS
InChi Key: SBRLITLVSMPPFI-UHFFFAOYSA-N
SMILES: ClCCCSC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results