accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4916-57-8

NN

CAS RN 4916-57-8

IUPAC Name: 4-[2-(pyridin-4-yl)ethyl]pyridine
Synonyms: 4-[2-(pyridin-4-yl)ethyl]pyridine
Molecular Weight (g/mol): 184.24
Molecular Formula: C12H12N2
InChi Key: DQRKTVIJNCVZAX-UHFFFAOYSA-N
SMILES: C(CC1=CC=NC=C1)C1=CC=NC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results