accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 492-37-5

IUPAC Name: 2-phenylpropanoic acid
Synonyms: 2-phenylpropionic acid (R)-isomer hydratropic acid α-methylbenzeneacetic acid hydratropic acid, (R)-isomer hydratropasaeure hydratropic acid, (S)-isomer (r)-hydratropic acid (s)-isomer hydratropic acid (+-)-hydratropasaeure (S)-isomer hydratropic acid
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: YPGCWEMNNLXISK-UHFFFAOYNA-N
SMILES: CC(C(O)=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (5)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (5)
  • (2)
  • (3)
  • (3)