accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4923-84-6

IUPAC Name: tert-butylphosphonate
Synonyms: tert-butylphosphonate
Molecular Weight (g/mol): 136.09
Molecular Formula: C4H9O3P
InChi Key: OGDSVONAYZTTDA-UHFFFAOYSA-L
SMILES: CC(C)(C)P([O-])([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results