accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4928-88-5

IUPAC Name: methyl 1H-1,2,4-triazole-5-carboxylate
Synonyms: methyl 2H-1,2,4-triazole-3-carboxylate
Molecular Weight (g/mol): 127.1
Molecular Formula: C4H5N3O2
InChi Key: QMPFMODFBNEYJH-UHFFFAOYSA-N
SMILES: COC(=O)C1=NC=NN1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results