CAS RN 493-52-7
Thermo Scientific Chemicals Methyl Red, pure, indicator
CAS: 493-52-7 Molecular Formula: C15H15N3O2 Molecular Weight (g/mol): 269.304 InChI Key: CEQFOVLGLXCDCX-UHFFFAOYSA-N Synonym: Acid Red 2,2-[4-(Dimethylamino)phenylazo]benzoic acid,C.I. 13020 PubChem CID: 10303 IUPAC Name: 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
Thermo Scientific Chemicals Methyl Red, ACS reagent
CAS: 493-52-7 Molecular Formula: C15H15N3O2 Molecular Weight (g/mol): 269.304 MDL Number: MFCD00002425 InChI Key: CEQFOVLGLXCDCX-UHFFFAOYSA-N Synonym: Acid Red 2,2-[4-(Dimethylamino)phenylazo]benzoic acid,C.I. 13020 PubChem CID: 10303 IUPAC Name: 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
Thermo Scientific Chemicals Methyl Red
CAS: 493-52-7 Molecular Formula: C15H15N3O2 Molecular Weight (g/mol): 269.304 MDL Number: MFCD00002425 InChI Key: CEQFOVLGLXCDCX-UHFFFAOYSA-N Synonym: C.I. 13020 PubChem CID: 10303 IUPAC Name: 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
Thermo Scientific Chemicals Methyl Red, ACS
CAS: 493-52-7 Molecular Formula: C15H15N3O2 Molecular Weight (g/mol): 269.304 MDL Number: MFCD00002425 InChI Key: CEQFOVLGLXCDCX-UHFFFAOYSA-N Synonym: C.I. 13020 PubChem CID: 10303 IUPAC Name: 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
Methyl red, For ACS analysis, MP Biomedicals™
CAS: 493-52-7 Molecular Formula: C15H15N3O2 Molecular Weight (g/mol): 269.304 InChI Key: CEQFOVLGLXCDCX-UHFFFAOYSA-N Synonym: 2-(4-Dimethylaminophenylazo)benzoic acid,4-Dimethylaminoazobenzene-2'-carboxylic acid PubChem CID: 10303 IUPAC Name: 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
Methyl Red Solution, EP Grade, Reagecon™
Turns red in acidic solutions and used in the MR test to indentify the presence of bacteria. Methyl Red Solution, EP Grade, Reagecon™ is a ready-to-use solution with 50mg methyl red per 100mL. It is manufactured according to Eur. Pharm. Chapter 4 (4.1.1) and prepared in an ISO 9001 facility.
Methyl Red, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Methyl Red, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Methyl Red, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Methyl Red Indicator, Reag. Ph. Eur., Solstice
CAS: 493-52-7 Molecular Formula: C15H15N3O2 Molecular Weight (g/mol): 269.304 MDL Number: MFCD00002425 InChI Key: CEQFOVLGLXCDCX-UHFFFAOYSA-N Synonym: Acid Red 2 PubChem CID: 10303 IUPAC Name: 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
Methyl Red, Pure, Spirit Soluble, C.I.13020, Indicator Grade, Fisher Chemical™
CAS: 493-52-7 Molecular Formula: C15H15N3O2 Molecular Weight (g/mol): 269.304 MDL Number: 2425 InChI Key: CEQFOVLGLXCDCX-UHFFFAOYSA-N PubChem CID: 10303 IUPAC Name: 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
Methyl Red, Pure, Water Soluble, Indicator, Fisher Chemical™
CAS: 493-52-7 Molecular Formula: C15H15N3O2 Molecular Weight (g/mol): 269.304 MDL Number: 2425 InChI Key: CEQFOVLGLXCDCX-UHFFFAOYSA-N PubChem CID: 10303 IUPAC Name: 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O