accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OO

CAS RN 4942-47-6

IUPAC Name: 2-(adamantan-1-yl)acetate
Synonyms: 2-(adamantan-1-yl)acetate
Molecular Weight (g/mol): 193.27
Molecular Formula: C12H17O2
InChi Key: AOTQGWFNFTVXNQ-UHFFFAOYSA-M
SMILES: [O-]C(=O)CC12CC3CC(CC(C3)C1)C2
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)