CAS RN 4944-60-9
IUPAC Name:
9-(acetyloxy)nonyl acetate
Synonyms:
nonanediol, diacetate
Molecular Weight (g/mol):
244.33
Molecular Formula:
C13H24O4
InChi Key:
JAEVCKMMGIFOCL-UHFFFAOYSA-N
SMILES:
CC(=O)OCCCCCCCCCOC(C)=O
1
–
1
of
1
results
1
1,9-Diacetoxynonane, TRC
CAS: 4944-60-9 Molecular Formula: C13H24O4 Molecular Weight (g/mol): 244.33 IUPAC Name: 9-acetyloxynonyl acetate SMILES: CC(=O)OCCCCCCCCCOC(=O)C