accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 49540-00-3

IUPAC Name: mercury(2+) ditrifluoromethanesulfonate
Synonyms: mercury(2+) ditriflate
Molecular Weight (g/mol): 498.72
Molecular Formula: C2F6HgO6S2
InChi Key: BPVYMDMPLCOQPJ-UHFFFAOYSA-L
SMILES: [Hg++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)