accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 496-11-7

IUPAC Name: 2,3-dihydro-1H-indene
Synonyms: indan hydrindene hydrindonaphthene 1,2-hydrindene
Molecular Weight (g/mol): 118.18
Molecular Formula: C9H10
InChi Key: PQNFLJBBNBOBRQ-UHFFFAOYSA-N
SMILES: C1CC2=CC=CC=C2C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (5)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)
  • (3)
  • (2)
  • (3)