CAS RN 496-74-2
IUPAC Name:
4-methylbenzene-1,2-dithiol
Synonyms:
1,2-benzenedithiol, 4-methyl-
Molecular Weight (g/mol):
156.26
Molecular Formula:
C7H8S2
InChi Key:
NIAAGQAEVGMHPM-UHFFFAOYSA-N
SMILES:
CC1=CC=C(S)C(S)=C1
1
–
1
of
1
results
1
Promotions Available
3,4-Dimercaptotoluene, 95%
3,4-Dimercaptotoluene, 95%
CAS: 496-74-2 Molecular Formula: C7H8S2 Molecular Weight (g/mol): 156.27 MDL Number: MFCD00004844 InChI Key: NIAAGQAEVGMHPM-UHFFFAOYSA-N Synonym: toluene-3,4-dithiol,dithiol,1,2-benzenedithiol, 4-methyl,3,4-dimercaptotoluene,3,4-dimercaptotoluol,1,2-dimercapto-4-methylbenzene,4-methyl-1,2-benzenedithiol,3,4-toluenedithiol,o-toluenesulfonylamide,usaf b-59 PubChem CID: 10334 IUPAC Name: 4-methylbenzene-1,2-dithiol SMILES: CC1=CC(=C(C=C1)S)S