accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 496-76-4

IUPAC Name: 5-hydroxy-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 5-hydroxyuracil isobarbituric acid isobarbituric acid (van) (8CI)
Molecular Weight (g/mol): 128.09
Molecular Formula: C4H4N2O3
InChi Key: OFJNVANOCZHTMW-UHFFFAOYSA-N
SMILES: OC1=CNC(=O)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results