accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNHSNH2CH3

CAS RN 49600-35-3

IUPAC Name: (4,6-dimethylpyridin-2-yl)thiourea
Synonyms: 4,6-dimethylpyridin-2-ylthiourea
Molecular Weight (g/mol): 181.26
Molecular Formula: C8H11N3S
InChi Key: ZXQTZSMIBLLDGW-UHFFFAOYSA-N
SMILES: CC1=CC(C)=NC(NC(N)=S)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results