accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4965-36-0

BrN

CAS RN 4965-36-0

IUPAC Name: 7-bromoquinoline
Synonyms: 7-bromoquinoline
Molecular Weight (g/mol): 208.06
Molecular Formula: C9H6BrN
InChi Key: XYBSZCUHOLWQQU-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C=CC=NC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results