CAS RN 496835-35-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methisosildenafil, TRC
CAS: 496835-35-9 Molecular Formula: C23 H32 N6 O4 S Molecular Weight (g/mol): 488.6 Synonym: 7H-Pyrazolo[4,3-d]pyrimidin-7-one, 5-[5-[[(3R,5S)-3,5-dimethyl-1-piperazinyl]sulfonyl]-2-ethoxyphenyl]-1,6-dihydro-1-methyl-3-propyl-, rel-,Piperazine, 1-[[3-(4,7-dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]-3,5-dimethyl-, (3R,5S)-rel- (9CI),Aildenafil,Methisosildenafil IUPAC Name: 5-[5-[(3S,5R)-3,5-dimethylpiperazin-1-yl]sulfonyl-2-ethoxyphenyl]-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-7-one SMILES: CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)N4C[C@@H](C)N[C@@H](C)C4