accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CH3CCH3NHSOONOO

CAS RN 49690-09-7

IUPAC Name: N-tert-butyl-4-nitrobenzene-1-sulfonamide
Synonyms: N-tert-butyl-4-nitrobenzenesulfonamide
Molecular Weight (g/mol): 258.29
Molecular Formula: C10H14N2O4S
InChi Key: YVSUQTZCHSBFEN-UHFFFAOYSA-N
SMILES: CC(C)(C)NS(=O)(=O)C1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results