CAS RN 49773-24-2
IUPAC Name:
ethyl 2-methyl-2-(methylsulfanyl)propanoate
Synonyms:
ethyl 2-methyl-2-(methylsulfanyl)propanoate
Molecular Weight (g/mol):
162.25
Molecular Formula:
C7H14O2S
InChi Key:
JLURSSXMQSLZQL-UHFFFAOYSA-N
SMILES:
CCOC(=O)C(C)(C)SC
1
–
1
of
1
results
1
Ethyl 2-methyl-2-(methylthio)propionate, 98%, Thermo Scientific™
CAS: 49773-24-2 Molecular Formula: C7H14O2S Molecular Weight (g/mol): 162.247 MDL Number: MFCD00053099 InChI Key: JLURSSXMQSLZQL-UHFFFAOYSA-N Synonym: ethyl 2-methyl-2-methylthio propionate,propanoic acid, 2-methyl-2-methylthio-, ethyl ester,ethyl 2-methyl-2-methylthio propanoate,ethyl 2-methyl-2-methylsulfanyl propanoate,acmc-20al0x,ethyl 2-methylthio isobutyrate,2-methyl-2-methylthio propionic acid ethyl ester,2-methyl-2-methylthio-propanoic acid ethyl ester PubChem CID: 3016499 IUPAC Name: ethyl 2-methyl-2-methylsulfanylpropanoate SMILES: CCOC(=O)C(C)(C)SC