accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 498-23-7

IUPAC Name: (2Z)-2-methylbut-2-enedioic acid
Synonyms: citraconic acid kyselina mesakonova mesaconate 2-methylfumaric acid methylfumaric acid
Molecular Weight (g/mol): 130.1
Molecular Formula: C5H6O4
InChi Key: HNEGQIOMVPPMNR-IHWYPQMZSA-N
SMILES: C\C(=C\C(O)=O)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)