accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 498-66-8

IUPAC Name: bicyclo[2.2.1]hept-2-ene
Synonyms: norbornene bicyclo[2.2.1]-2-heptene
Molecular Weight (g/mol): 94.16
Molecular Formula: C7H10
InChi Key: JFNLZVQOOSMTJK-UHFFFAOYNA-N
SMILES: C1CC2CC1C=C2
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (6)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)