accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOHN

CAS RN 498-95-3

IUPAC Name: (3R)-piperidin-1-ium-3-carboxylate
Synonyms: (3R)-piperidine-3-carboxylate (R)-nipecotic acid zwitterion (3R)-nipecotic acid zwitterion
Molecular Weight (g/mol): 129.16
Molecular Formula: C6H11NO2
InChi Key: XJLSEXAGTJCILF-RXMQYKEDSA-N
SMILES: [O-]C(=O)[C@@H]1CCC[NH2+]C1
Quantity 
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)