accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3HNOF

CAS RN 49834-29-9

IUPAC Name: N-tert-butyl-4-fluorobenzamide
Synonyms: N-tert-butyl-4-fluorobenzamide
Molecular Weight (g/mol): 195.24
Molecular Formula: C11H14FNO
InChi Key: ZMYTVKMZSNODPU-UHFFFAOYSA-N
SMILES: CC(C)(C)NC(=O)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results