accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaHC

CAS RN 4984-82-1

IUPAC Name: sodium cyclopenta-2,4-dien-1-ide
Synonyms: sodium cyclopentadienide
Molecular Weight (g/mol): 88.09
Molecular Formula: C5H5Na
InChi Key: OHUVHDUNQKJDKW-UHFFFAOYSA-N
SMILES: [Na+].[CH-]1C=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results