accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 499-40-1

HO(R)O(S)O(R)OOH(R)OH(S)OH(S)HO(R)OH(S)HO(S)HO

CAS RN 499-40-1

IUPAC Name: (3R,4S,5S,6R)-6-({[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxane-2,3,4,5-tetrol
Synonyms: isomaltose brachiose
Molecular Weight (g/mol): 342.3
Molecular Formula: C12H22O11
InChi Key: DLRVVLDZNNYCBX-RTPHMHGBSA-N
SMILES: OC[C@H]1O[C@H](OC[C@H]2OC(O)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)