accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHONOHO

CAS RN 499-81-0

IUPAC Name: pyridine-3,5-dicarboxylic acid
Synonyms: 3,5-pyridinedicarboxylic acid dinicotinic acid
Molecular Weight (g/mol): 167.12
Molecular Formula: C7H5NO4
InChi Key: MPFLRYZEEAQMLQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CN=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)