accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 499770-88-6

IUPAC Name: 5-(benzyloxy)-2-bromo-4-methylaniline
Synonyms: 5-(benzyloxy)-2-bromo-4-methylaniline
Molecular Weight (g/mol): 292.18
Molecular Formula: C14H14BrNO
InChi Key: FEMSDHJSFUHYSH-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=C(N)C=C1OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results