accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClNH3

CAS RN 4998-76-9

IUPAC Name: cyclohexanaminium chloride
Synonyms: cyclohexylammonium ion chloride
Molecular Weight (g/mol): 135.64
Molecular Formula: C6H14ClN
InChi Key: ZJUGSKJHHWASAF-UHFFFAOYSA-N
SMILES: [Cl-].[NH3+]C1CCCCC1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results