CAS RN 50-01-1
Guanidine hydrochloride, 98%
CAS: 50-01-1 Molecular Formula: CH6ClN3 Molecular Weight (g/mol): 95.53 MDL Number: MFCD00013026 InChI Key: PJJJBBJSCAKJQF-UHFFFAOYSA-N Synonym: guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride PubChem CID: 5742 ChEBI: CHEBI:32735 IUPAC Name: guanidine;hydrochloride SMILES: C(=N)(N)N.Cl
Guanidine Hydrochloride (Colorless-to-White Crystals), Fisher BioReagents™
For isolation of RNA from cell extracts | CAS: 50-01-1 | CH6ClN3
Thermo Scientific Chemicals Guanidine hydrochloride, ultrapure, 99%
CAS: 50-01-1 Molecular Formula: CH6ClN3 Molecular Weight (g/mol): 95.53 MDL Number: MFCD00013026 InChI Key: PJJJBBJSCAKJQF-UHFFFAOYSA-N Synonym: guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride PubChem CID: 5742 ChEBI: CHEBI:32735 IUPAC Name: guanidine;hydrochloride SMILES: C(=N)(N)N.Cl
Thermo Scientific Chemicals Guanidine hydrochloride, Molecular Biology Grade
CAS: 50-01-1 Molecular Formula: CH6ClN3 Molecular Weight (g/mol): 95.53 MDL Number: MFCD00013026 InChI Key: PJJJBBJSCAKJQF-UHFFFAOYSA-N Synonym: guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride PubChem CID: 5742 ChEBI: CHEBI:32735 IUPAC Name: guanidine;hydrochloride SMILES: C(=N)(N)N.Cl
Thermo Scientific Chemicals Guanidine hydrochloride, 99+%
CAS: 50-01-1 Molecular Formula: CH6ClN3 Molecular Weight (g/mol): 95.53 MDL Number: MFCD00013026 InChI Key: PJJJBBJSCAKJQF-UHFFFAOYSA-N Synonym: guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride PubChem CID: 5742 ChEBI: CHEBI:32735 IUPAC Name: guanidine;hydrochloride SMILES: C(=N)(N)N.Cl
Guanidine Hydrochloride, >99.5%, Molecular Biology Grade
CAS: 50-01-1 Molecular Formula: CH6ClN3 Molecular Weight (g/mol): 95.53 MDL Number: MFCD00013026 InChI Key: PJJJBBJSCAKJQF-UHFFFAOYSA-N Synonym: guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride PubChem CID: 5742 ChEBI: CHEBI:32735 IUPAC Name: guanidine;hydrochloride SMILES: C(=N)(N)N.Cl
Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2)
CAS: 50-01-1 Molecular Formula: CH5N3·HCl Molecular Weight (g/mol): 95.53 InChI Key: PJJJBBJSCAKJQF-UHFFFAOYSA-N Synonym: guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride PubChem CID: 5742 ChEBI: CHEBI:32735 IUPAC Name: guanidine;hydrochloride SMILES: C(=N)(N)N.Cl
Guanidine hydrochloride, 99.5%, without anticaking agent
CAS: 50-01-1 Molecular Formula: CH5N3·HCl Molecular Weight (g/mol): 95.53 MDL Number: MFCD00013026 InChI Key: PJJJBBJSCAKJQF-UHFFFAOYSA-N Synonym: guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride PubChem CID: 5742 ChEBI: CHEBI:32735 IUPAC Name: guanidine;hydrochloride SMILES: C(=N)(N)N.Cl
Guanidine Hydrochloride, Ultra Pure grade, MP Biomedicals
CAS: 50-01-1 Molecular Formula: CH6ClN3 Molecular Weight (g/mol): 95.53 MDL Number: MFCD00013026 InChI Key: PJJJBBJSCAKJQF-UHFFFAOYSA-N Synonym: guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride PubChem CID: 5742 ChEBI: CHEBI:32735 IUPAC Name: guanidine;hydrochloride SMILES: C(=N)(N)N.Cl
Guanidine Hydrochloride, ≥99.5%, (Crystalline/Ultra Pure), MP Biomedicals
CAS: 50-01-1 Molecular Formula: CH6ClN3 Molecular Weight (g/mol): 95.53 InChI Key: PJJJBBJSCAKJQF-UHFFFAOYSA-N Synonym: guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride PubChem CID: 5742 ChEBI: CHEBI:32735 IUPAC Name: guanidine;hydrochloride SMILES: C(=N)(N)N.Cl
Guanidine Hydrochloride, 99+%, MP Biomedicals
CAS: 50-01-1 Molecular Formula: CH6ClN3 Molecular Weight (g/mol): 95.53 InChI Key: PJJJBBJSCAKJQF-UHFFFAOYSA-N Synonym: guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride PubChem CID: 5742 ChEBI: CHEBI:32735 IUPAC Name: guanidine;hydrochloride SMILES: C(=N)(N)N.Cl
Guanidine Hydrochloride, (Crystalline, solution may be cloudy and yellow/Practical Grade), MP Biomedicals
CAS: 50-01-1 Molecular Formula: CH6ClN3 Molecular Weight (g/mol): 95.53 InChI Key: PJJJBBJSCAKJQF-UHFFFAOYSA-N Synonym: guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride PubChem CID: 5742 ChEBI: CHEBI:32735 IUPAC Name: guanidine;hydrochloride SMILES: C(=N)(N)N.Cl
Guanidine Hydrochloride, ∽99%, (Crystalline/Optical Grade), MP Biomedicals
CAS: 50-01-1 Molecular Formula: CH6ClN3 Molecular Weight (g/mol): 95.53 InChI Key: PJJJBBJSCAKJQF-UHFFFAOYSA-N Synonym: guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride PubChem CID: 5742 ChEBI: CHEBI:32735 IUPAC Name: guanidine;hydrochloride SMILES: C(=N)(N)N.Cl
- 1
- 2