CAS RN 50-23-7
Hydrocortisone, 98%
CAS: 50-23-7 Molecular Formula: C21H30O5 MDL Number: MFCD00011654 InChI Key: JYGXADMDTFJGBT-VWUMJDOOSA-N Synonym: hydrocortisone,cortisol,acticort,cetacort,cortef,17-hydroxycorticosterone,hydrocortisyl,hydrasson,cobadex,cortril PubChem CID: 5754 ChEBI: CHEBI:17650
Thermo Scientific Chemicals Hydrocortisone, 98%
CAS: 50-23-7 Molecular Formula: C21H30O5 Molecular Weight (g/mol): 362.47 MDL Number: MFCD00011654 InChI Key: JYGXADMDTFJGBT-VWUMJDOOSA-N Synonym: hydrocortisone,cortisol,acticort,cetacort,cortef,17-hydroxycorticosterone,hydrocortisyl,hydrasson,cobadex,cortril PubChem CID: 5754 ChEBI: CHEBI:17650 IUPAC Name: (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one SMILES: CC12CCC(=O)C=C1CCC3C2C(CC4(C3CCC4(C(=O)CO)O)C)O
Hydrocortisone, ≥97%, MP Biomedicals™
CAS: 50-23-7 Molecular Formula: C21H30O5 Molecular Weight (g/mol): 362.466 MDL Number: MFCD00011654 InChI Key: JYGXADMDTFJGBT-VWUMJDOOSA-N Synonym: hydrocortisone,cortisol,acticort,cetacort,cortef,17-hydroxycorticosterone,hydrocortisyl,hydrasson,cobadex,cortril PubChem CID: 5754 ChEBI: CHEBI:17650 IUPAC Name: (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one SMILES: CC12CCC(=O)C=C1CCC3C2C(CC4(C3CCC4(C(=O)CO)O)C)O
R&D Systems™ Hydrocortisone
Hydrocortisone (Cortisol), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Hydrocortisone (Cortisol), TRC
CAS: 50-23-7 Molecular Formula: C21 H30 O5 Molecular Weight (g/mol): 362.46 Synonym: Hydrocortisone,Cortisol,11beta,17,21-Trihydroxy-pregn-4-ene-3,20-dione,Hydrocortisone Hydrogen Succinate Imp. A (EP),Prednisolone Imp. A (EP),Hydrocortisone Acetate Imp. A (Pharmeuropa),11β,17,21-Trihydroxyprogesterone,11β,17α,21-Trihydroxypregn-4-ene-3,20-dione,11β-Hydroxycortisone,17-Hydroxycorticosterone,17α-Hydroxycorticosterone,4-Pregnene-11β,17α,21-triol-3,20-dione,Acticort,Aeroseb HC,Ala-Cort,Anflam,Anti-inflammatory hormone,CCN 90306A,CaldeCort Spray,Cetacort,Cobadex,Cort-Dome,Cortanal,Cortef,Cortenema,Corticreme,Cortifan,Cortiment,Cortispray,Cortonema,Cortril,Dermacort,Dermocortal,Dermolate,Dihydrocostisone,Dioderm,Domolene-HC,Efcorbin,Efcortelan,Eldecort,Epiderm H,Esiderm H,Evacort,Ficortril,Genacort,HC,Heb-Cort,Hidro-Colisona,Hycort,Hycortol,Hycortole,Hydracort,Hydrasson,Hydro-Adreson,Hydrocort,Hydrocortal,Hydrocorticosterone,Hydrocortisone,Hydrocortisyl,Hydrocortone,Hydroskin,Hytone,Hytone lotion,Incortin-H,Kendall's compound F,Lacticare HC,Medicort,Mildison,NSC 10483,Nutracort,Optef,Otosone-F,Penecort,Permicort,Polcort H,Reichstein's substance M,Scheroson F,Signef,Synacort,Tarcortin,Texacort,Texacort lotion 25,Timocort,Topicort,Traumaide,U 1851,Zenoxone IUPAC Name: (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one SMILES: C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)CO)[C@@]4(C)C[C@H](O)[C@H]23
Cortisol, Lipomed
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Cortisol, Lipomed
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Cortisol, Lipomed
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Hydrocortisone (Cortisol), LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
Cortisol, 1mg/ml in Methanol, Lipomed
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.