accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 50-32-8

IUPAC Name: pentacyclo[10.6.2.0²,⁷.0⁹,¹⁹.0¹⁶,²⁰]icosa-1(18),2,4,6,8,10,12,14,16,19-decaene
Synonyms: benzopyrene benzo(d,e,f)chrysene 3,4-benzopirene 3,4-benzo(a)pyrene 3,4 benzopyrene 3,4-benzpyren 3,4 benzpyrene BaP benzo(A)pyrene 6,7-benzopyrene
Molecular Weight (g/mol): 252.32
Molecular Formula: C20H12
InChi Key: FMMWHPNWAFZXNH-UHFFFAOYSA-N
SMILES: C1=CC=C2C(=C1)C=C1C=CC3=CC=CC4=CC=C2C1=C34
Molecular Weight (g/mol) 
  • (3)
  • (7)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Solvent or Matrix 
  • (2)
  • (2)
  • (1)
Concentration 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (7)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)