accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOOOHN

CAS RN 50-35-1

IUPAC Name: 2-(2,6-dioxopiperidin-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: thalidomide calmore asmadion distoval profarmil hippuzon n-phthalylglutamic acid imide neurodyn kevadon ( -)-thalidomide
Molecular Weight (g/mol): 258.23
Molecular Formula: C13H10N2O4
InChi Key: UEJJHQNACJXSKW-UHFFFAOYNA-N
SMILES: O=C1N(C2CCC(=O)NC2=O)C(=O)C2=CC=CC=C12
Quantity 
  • (1)
  • (4)
  • (1)
Physical Form 
  • (2)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)