CAS RN 50-62-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hexobendine Dihydrochloride, TRC
CAS: 50-62-4 Molecular Formula: C30 H44 N2 O10 . 2 Cl H Molecular Weight (g/mol): 665.6 Synonym: 3,4,5-Trimethoxybenzoic Acid 1,2-Ethanediylbis[(methylimino)-3,1-propanediyl] Ester Dihydrochloride,3,3'-[Ethylenebis(methylimino)]di-1-propanol Bis(3,4,5-trimethoxybenzoate) (Ester) Dihydrochloride,Andiamine,Hexobendin Hydrochloride,Reoxyl IUPAC Name: 3-[methyl-[2-[methyl-[3-(3,4,5-trimethoxybenzoyl)oxypropyl]amino]ethyl]amino]propyl 3,4,5-trimethoxybenzoate;dihydrochloride SMILES: Cl.Cl.COc1cc(cc(OC)c1OC)C(=O)OCCCN(C)CCN(C)CCCOC(=O)c2cc(OC)c(OC)c(OC)c2