accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3HNOHNOCH3OCH3OONHCH3OOH3CCH3NH3CONH3CONONHOCH3CH3NNH2CH3OOCH3CH3NH3CONH3CONO

CAS RN 50-76-0

IUPAC Name: 2-amino-4,6-dimethyl-3-oxo-N1,N9-bis[2,5,9-trimethyl-1,4,7,11,14-pentaoxo-6,13-bis(propan-2-yl)-hexadecahydro-1H-pyrrolo[2,1-i]1-oxa-4,7,10,13-tetraazacyclohexadecan-10-yl]-3H-phenoxazine-1,9-dicarboxamide
Synonyms: dactinomycin dilactone actinomycindioic D acid merck sharp & dohme brand of dactinomycin cosmegen cosmegen lyovac ac-de lyovac cosmegen ccris 9 hbf 386 meractinomycin actinomycin I(sub 1)
Molecular Weight (g/mol): 1255.44
Molecular Formula: C62H86N12O16
InChi Key: RJURFGZVJUQBHK-UHFFFAOYNA-N
SMILES: CC(C)C1NC(=O)C(NC(=O)C2=C3N=C4C(OC3=C(C)C=C2)=C(C)C(=O)C(N)=C4C(=O)NC2C(C)OC(=O)C(C(C)C)N(C)C(=O)CN(C)C(=O)C3CCCN3C(=O)C(NC2=O)C(C)C)C(C)OC(=O)C(C(C)C)N(C)C(=O)CN(C)C(=O)C2CCCN2C1=O
Molecular Weight (g/mol) 
  • (3)
  • (8)
Quantity 
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (4)
  • (2)
Physical Form 
  • (4)
  • (2)
  • (2)
Type 
  • (8)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (8)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (8)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)