accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 50-79-3

IUPAC Name: 2,5-dichlorobenzoic acid
Synonyms: 2,5-dichlorobenzoic acid
Molecular Weight (g/mol): 191.01
Molecular Formula: C7H4Cl2O2
InChi Key: QVTQYSFCFOGITD-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Cl)=CC=C1Cl
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results