accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 50-89-5

IUPAC Name: 1-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: TdR thymidine (substance) thymine 2'-deoxyriboside TDR deoxyribothymidine thymine-2-desoxyriboside 5-methyldeoxyuridine thymine 2-desoxyriboside 5-methyldeoxyurindine dthyd
Molecular Weight (g/mol): 242.23
Molecular Formula: C10H14N2O5
InChi Key: IQFYYKKMVGJFEH-UHFFFAOYNA-N
SMILES: CC1=CN(C2CC(O)C(CO)O2)C(=O)NC1=O
Quantity 
  • (1)
  • (1)
  • (3)
  • (4)
  • (3)
Percent Purity 
  • (4)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (4)
  • (6)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (3)
  • (4)
  • (3)
  • (3)
  • (3)
  • (7)