CAS RN 50-99-7
Thermo Scientific Chemicals D-(+)-Glucose, anhydrous, 99%
CAS: 50-99-7 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.156 MDL Number: MFCD00063774 InChI Key: GZCGUPFRVQAUEE-SLPGGIOYSA-N Synonym: 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal,d +-glucose,aldehydo-d-glucose,glucose, anhydrous,dextrose, anhydrous,d-glucose, anhydrous,glucosteril,anhydrous dextrose,dextrose solution,d-glucose in linear form PubChem CID: 107526 ChEBI: CHEBI:42758 IUPAC Name: (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal SMILES: C(C(C(C(C(C=O)O)O)O)O)O
D-(+)-Glucose, 1M aq. soln., sterile-filtered
CAS: 50-99-7 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.156 MDL Number: MFCD00148912 InChI Key: GZCGUPFRVQAUEE-SLPGGIOYSA-N Synonym: 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal,d +-glucose,aldehydo-d-glucose,glucose, anhydrous,dextrose, anhydrous,d-glucose, anhydrous,glucosteril,anhydrous dextrose,dextrose solution,d-glucose in linear form PubChem CID: 107526 ChEBI: CHEBI:42758 IUPAC Name: (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal SMILES: C(C(C(C(C(C=O)O)O)O)O)O
D(+)-Glucose Anhydrous, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™
CAS: 50-99-7 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.156 MDL Number: 148912 InChI Key: GZCGUPFRVQAUEE-SLPGGIOYSA-N Synonym: 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal,d +-glucose,aldehydo-d-glucose,glucose, anhydrous,dextrose, anhydrous,d-glucose, anhydrous,glucosteril,anhydrous dextrose,dextrose solution,d-glucose in linear form PubChem CID: 107526 ChEBI: CHEBI:42758 IUPAC Name: (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal SMILES: C(C(C(C(C(C=O)O)O)O)O)O
D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP
CAS: 50-99-7 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.16 InChI Key: GZCGUPFRVQAUEE-SLPGGIOYSA-N Synonym: 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal,d +-glucose,aldehydo-d-glucose,glucose, anhydrous,dextrose, anhydrous,d-glucose, anhydrous,glucosteril,anhydrous dextrose,dextrose solution,d-glucose in linear form PubChem CID: 107526 ChEBI: CHEBI:42758 IUPAC Name: (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal SMILES: C(C(C(C(C(C=O)O)O)O)O)O
D(+)-Glucose Anhydrous, Extra Pure, SLR, Fisher Chemical™
CAS: 50-99-7 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.156 MDL Number: 148912 InChI Key: GZCGUPFRVQAUEE-SLPGGIOYSA-N Synonym: 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal,d +-glucose,aldehydo-d-glucose,glucose, anhydrous,dextrose, anhydrous,d-glucose, anhydrous,glucosteril,anhydrous dextrose,dextrose solution,d-glucose in linear form PubChem CID: 107526 ChEBI: CHEBI:42758 IUPAC Name: (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal SMILES: C(C(C(C(C(C=O)O)O)O)O)O
D(+)-Glucose, ACS Reagent, anhydrous
CAS: 50-99-7 MDL Number: MFCD00063684 InChI Key: GZCGUPFRVQAUEE-SLPGGIOYSA-N Synonym: 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal,d +-glucose,aldehydo-d-glucose,glucose, anhydrous,dextrose, anhydrous,d-glucose, anhydrous,glucosteril,anhydrous dextrose,dextrose solution,d-glucose in linear form PubChem CID: 107526 ChEBI: CHEBI:42758 IUPAC Name: (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal SMILES: C(C(C(C(C(C=O)O)O)O)O)O
Dextrose Anhydrous (Crystalline Granules/Molecular Biology), Fisher BioReagents
CAS: 50-99-7 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.156 InChI Key: GZCGUPFRVQAUEE-SLPGGIOYSA-N Synonym: 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal,d +-glucose,aldehydo-d-glucose,glucose, anhydrous,dextrose, anhydrous,d-glucose, anhydrous,glucosteril,anhydrous dextrose,dextrose solution,d-glucose in linear form PubChem CID: 107526 ChEBI: CHEBI:42758 IUPAC Name: (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal SMILES: C(C(C(C(C(C=O)O)O)O)O)O
D-(+)-Dextrose, Anhydrous
CAS: 50-99-7 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.156 MDL Number: MFCD00063774 InChI Key: GZCGUPFRVQAUEE-SLPGGIOYSA-N Synonym: 2 r,3 s,4 r,5 r-2,3,4,5,6-pentahydroxyhexanal,d +-glucose,aldehydo-d-glucose,glucose, anhydrous,dextrose, anhydrous,d-glucose, anhydrous,glucosteril,anhydrous dextrose,dextrose solution,d-glucose in linear form PubChem CID: 107526 ChEBI: CHEBI:42758 IUPAC Name: (2 R,3 S,4 R,5 R)-2,3,4,5,6-pentahydroxyhexanal SMILES: C(C(C(C(C(C=O)O)O)O)O)O
D-(+)-Dextrose, (Crystalline), MP Biomedicals
CAS: 50-99-7 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.156 InChI Key: GZCGUPFRVQAUEE-SLPGGIOYSA-N Synonym: 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal,d +-glucose,aldehydo-d-glucose,glucose, anhydrous,dextrose, anhydrous,d-glucose, anhydrous,glucosteril,anhydrous dextrose,dextrose solution,d-glucose in linear form PubChem CID: 107526 ChEBI: CHEBI:42758 IUPAC Name: (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal SMILES: C(C(C(C(C(C=O)O)O)O)O)O
D-(+)-Glucose, Molecular biology reagent grade, MP Biomedicals™
CAS: 50-99-7 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.156 InChI Key: GZCGUPFRVQAUEE-SLPGGIOYSA-N Synonym: 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal,d +-glucose,aldehydo-d-glucose,glucose, anhydrous,dextrose, anhydrous,d-glucose, anhydrous,glucosteril,anhydrous dextrose,dextrose solution,d-glucose in linear form PubChem CID: 107526 ChEBI: CHEBI:42758 IUPAC Name: (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal SMILES: C(C(C(C(C(C=O)O)O)O)O)O
D-(+)-Glucose, anhydrous, For ACS analysis, MP Biomedicals™
CAS: 50-99-7 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.156 InChI Key: GZCGUPFRVQAUEE-SLPGGIOYSA-N Synonym: 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal,d +-glucose,aldehydo-d-glucose,glucose, anhydrous,dextrose, anhydrous,d-glucose, anhydrous,glucosteril,anhydrous dextrose,dextrose solution,d-glucose in linear form PubChem CID: 107526 ChEBI: CHEBI:42758 IUPAC Name: (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal SMILES: C(C(C(C(C(C=O)O)O)O)O)O
D-(+)-Glucose, (Crystalline), MP Biomedicals
- 1
- 2