accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HBrNH2Br

CAS RN 5003-71-4

IUPAC Name: hydrogen 3-bromopropan-1-amine bromide
Synonyms: hydrogen 3-bromopropan-1-amine bromide
Molecular Weight (g/mol): 218.92
Molecular Formula: C3H9Br2N
InChi Key: PQIYSSSTRHVOBW-UHFFFAOYSA-N
SMILES: [H+].[Br-].NCCCBr
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results