accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClO

CAS RN 5006-22-4

IUPAC Name: cyclobutanecarbonyl chloride
Synonyms: cyclobutanecarbonyl chloride
Molecular Weight (g/mol): 118.56
Molecular Formula: C5H7ClO
InChi Key: JFWMYCVMQSLLOO-UHFFFAOYSA-N
SMILES: ClC(=O)C1CCC1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)